Magnesium chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Magnesium dichloride
M-Clarity™ quality level = MQ200
63068-1KG 63068-250G
SIAL63068-1KGEA 218 EUR
SIAL63068-1KG SIAL63068-250G
Magnesium chloride hexahydrate, Sigma-Aldrich®
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412
Melting Pt: 117
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684