1,2-Dibromoethane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
03505-100ML 03505-1L 03505-500ML
SIAL03505-100MLEA 34.5 EUR
SIAL03505-100ML SIAL03505-1L SIAL03505-500ML
1,2-Dibromoethane, Sigma-Aldrich®
1,2-Dibromoethane
Formula: BrH₂CCH₂Br
MW: 187.86 g/mol
Boiling Pt: 131.7
Melting Pt: 10
Density: 2.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000233
CAS Number: 106-93-4
EINECS: 203-444-5
UN: 1605
ADR: 6.1 I
Merck Index: 13,03830