1-Bromo-4-chlorobutane 99%
Supplier: Thermo Fisher Scientific
Warning
106840250 106841000
ACRO106840250EA 53.7 EUR
ACRO106840250 ACRO106841000
1-Bromo-4-chlorobutane 99%
1-Bromo-4-chlorobutane
Formula: BrH₂C(CH₂)₂CH₂Cl
Boiling Pt: 80…82 °C (30 mmHg)
Density: 1.48 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00001010
CAS Number: 6940-78-9
EINECS: 230-089-3
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4865 to 1.4885 (20°C, 589 nm)
GC ≥98.5 %