Bromotrichloromethane, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
B82251-100G
B82251-500G
SIALB82251-100GEA
49.7
EUR
SIALB82251-100G
SIALB82251-500G
Bromotrichloromethane, Sigma-Aldrich®
Bromotrichloromethane
Formula:
BrCl₃C MW: 198.27 g/mol Boiling Pt: 105 Melting Pt: –6 °C Density: 2.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000783 CAS Number: 75-62-7 EINECS: 200-886-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...