Tributylallyltin 97%

Supplier: Thermo Fisher Scientific
Danger

265550050 265550250
ACRO265550050EA 57.1 EUR
ACRO265550050 ACRO265550250
Tributylallyltin 97%
Tributylallyltin
Formula: C₁₅H₃₂Sn
Boiling Pt: 88…92 °C (.2 mmHg)
Density: 1.068 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4870 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR