(3,3,3-Trifluoropropyl)trimethoxysilane, Sigma-Aldrich®
Supplier: Merck
Synonyms: Trimethoxy(3,3,3-trifluoropropyl)silane
91877-25ML 91877-5ML
SIAL91877-25MLEA 305 EUR
SIAL91877-25ML SIAL91877-5ML
(3,3,3-Trifluoropropyl)trimethoxysilane, Sigma-Aldrich®
(3,3,3-Trifluoropropyl)trimethoxysilane
Formula: F₃CCH₂CH₂Si(OCH₃)₃
MW: 218.25 g/mol
Boiling Pt: 144
Density: 1.137 g/cm³ (20 °C)
MDL Number: MFCD00039266
CAS Number: 429-60-7
EINECS: 207-059-3