(4-Bromobutyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23040.09
B23040.18
B23040.09EA
44.8
EUR
B23040.09
B23040.18
(4-Bromobutyl)triphenylphosphonium bromide ≥98%
(4-Bromobutyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 7333-63-3
Molecular Formula: Br(CH2)4P(C6H5)3Br
Formula Weight: 478.22
Melting Point: 206-212°
Beilstein Reference: 3812291
Formula:
C₂₂H₂₃Br₂P MW: 478.21 g/mol Melting Pt: 206…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031636 CAS Number: 7333-63-3 |