(-)-1,4-Dithio-L-threitol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: L-Cleland's Reagent, L-threo-1,4-Dimercaptobutane-2,3-diol, (2R,3R)-1,4-Dimercaptobutane-2,3-diol, L-Dithiothreitol, L-DTT, [R-(R*,R*)]-1,4-Dimercapto-2,3-butandiol, (2R,3R)-(-)-1,4-Dimercapto-2,3-butanediol, (2R,3R)-(-)-2,3-Dihydroxy-1,4-butanedithiol
M-Clarity™ quality level = MQ100
D9760-1G D9760-500MG
SIALD9760-1GEA 306 EUR
SIALD9760-1G SIALD9760-500MG
(-)-1,4-Dithio-L-threitol, Sigma-Aldrich®
(-)-1,4-Dithio-L-threitol
Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00064305
CAS Number: 16096-97-2
EINECS: 240-263-0