2,3-Dimethyl-4-fluoroaniline ≥97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Fluoro-2,3-dimethylaniline, 4-Fluoro-2,3-xylidine, 3-Amino-6-fluoro-o-xylene
H31560.MD H31560.03
H31560.MDEA 79.1 EUR
H31560.MD H31560.03
2,3-Dimethyl-4-fluoroaniline ≥97%
2,3-Dimethyl-4-fluoroaniline
Purity: 97% CAS No.: 1737-68-4 Molecular Formula: C8H10FN Formula Weight: 139.17 Melting Point: 42-46° UN No.: 2811 MDL No.: MFCD04972870
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD04972870
CAS Number: 1737-68-4
UN: 2811
ADR: 6.1 III