2'-Hydroxy-5'-methylacetophenone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-(2-Hydroxy-5-methylphenyl)ethanone

H37601-10G H37601-50G
SIALH37601-10GEA 39.1 EUR
SIALH37601-10G SIALH37601-50G
2'-Hydroxy-5'-methylacetophenone, Sigma-Aldrich®
2'-Hydroxy-5'-methylacetophenone
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 83…87 °C (0.5 mmHg)
Melting Pt: 53
Density: 1.0797 g/cm³ (53 °C)
MDL Number: MFCD00002380
CAS Number: 1450-72-2
EINECS: 215-915-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR