3'-Fluoroacetophenone
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-(3-Fluorophenyl)ethanone, m-Fluoroacetophenone
SB01647.1
SB01647.10
MAYBSB01647.1EA
0
EUR
MAYBSB01647.1
MAYBSB01647.10
3'-Fluoroacetophenone
3'-Fluoroacetophenone
Formula:
C₈H₇FO Boiling Pt: 204 °C (1013 hPa) Melting Pt: –3 °C Density: 1,12 g/cm³ (20 °C) |
MDL Number:
MFCD00000338 CAS Number: 455-36-7 EINECS: 207-245-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...