1-Pentadecene ≥97%
Supplier: Thermo Fisher Scientific
H53516.06
H53516.14
H53516.06EA
80.9
EUR
H53516.06
H53516.14
1-Pentadecene ≥97%
1-Pentadecene
Purity: 97%
CAS No.: 13360-61-7
Molecular Formula: C15H30
Formula Weight: 210.40
Flash Point: 110°(230°F)
Refractive Index: 1.439
Melting Point: -4°
Boiling Point: 268-269°
Beilstein Reference: 1751011
MDL No.: MFCD00008987
Formula:
C₁₅H₃₀ Boiling Pt: 268…269 °C Melting Pt: –4 °C Density: 0.775 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008987 CAS Number: 13360-61-7 EINECS: 236-414-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...