2-Acetylfuran, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Furyl methyl ketone, 1-(Furan-2-yl)ethanone

M-Clarity™ quality level = MQ300

W316318-10KG-K W316318-1KG-K W316318-5KG-K
SIALW316318-10KG-KEA 727 EUR
SIALW316318-10KG-K SIALW316318-1KG-K SIALW316318-5KG-K
2-Acetylfuran, Sigma-Aldrich®
2-Acetylfuran
Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 183
Melting Pt: 29…33 °C
Density: 1.098 g/cm³ (20 °C)
MDL Number: MFCD00003242
CAS Number: 1192-62-7
EINECS: 214-757-1
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR