2-Vinylpyridine 97% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
158000050
158002500
158000010
ACRO158000050EA
31.8
EUR
ACRO158000050
ACRO158002500
ACRO158000010
2-Vinylpyridine 97% stabilised
2-Vinylpyridine
Formula:
C₇H₇N Boiling Pt: 159 °C (760 mmHg) Melting Pt: –50 °C Density: 0.9983 g/cm³ Flash Pt: 47 °C Storage Temperature: Freezer |
MDL Number:
MFCD00006355 CAS Number: 100-69-6 EINECS: 202-879-8 UN: 3073 ADR: 6.1,II |
Specification Test Results
Appearance | Clear brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5490 to 1.5510 (20°C, 589 nm) |
GC | ≥96.0 % |
Stabilizer | 0.1 % tert.-butylcatechol |
UV | at 400 nm A: ≤0.01 |
Color scale | ≤15 APHA |
Residue after evaporation | ≤0.01 % |
Heavy metals (as Pb) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...