2-Heptylfuran 97%
Supplier: Thermo Fisher Scientific
Warning
|
A10604.09
A10604.18
A10604.09EA
66.5
EUR
A10604.09
A10604.18
2-Heptylfuran 97%
2-Heptylfuran
Purity: 98%
CAS No.: 3777-71-7
Molecular Formula: C11H18O
Formula Weight: 166.27
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Refractive Index: 1.45
Boiling Point: 96-98°/30mm
MDL No.: MFCD00051820
Formula:
C₁₁H₁₈O Boiling Pt: 96…98 °C (30 mmHg) Density: 0.863 Flash Pt: 75 °C (167 °F) |
MDL Number:
MFCD00051820 CAS Number: 3777-71-7 EINECS: 223-236-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...