2-Hexylfuran ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H26698.03
H26698.06
H26698.14
H26698.03EA
40.1
EUR
H26698.03
H26698.06
H26698.14
2-Hexylfuran ≥97%
2-Hexylfuran
Purity: 97%
CAS No.: 3777-70-6
Molecular Formula: C10H16O
Formula Weight: 152.24
UN No.: 1993
MDL No.: MFCD00053134
Formula:
C₁₀H₁₆O Storage Temperature: Ambient |
MDL Number:
MFCD00053134 CAS Number: 3777-70-6 EINECS: 223-235-2 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...