Ethyl-2-methylacetoacetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
E35400-100G E35400-25G
SIALE35400-100GEA 136 EUR
SIALE35400-100G SIALE35400-25G
Ethyl-2-methylacetoacetate, Sigma-Aldrich®
Ethyl 2-methylacetoacetate
Formula: C₇H₁₂O₃
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.0238 g/cm³ (15.3 °C)
MDL Number: MFCD00009164
CAS Number: 609-14-3
EINECS: 210-179-9