2-Pentylfuran, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W331708-100G-K
W331708-1KG-K
W331708-25G-K
SIALW331708-100G-KEA
552
EUR
SIALW331708-100G-K
SIALW331708-1KG-K
SIALW331708-25G-K
2-Pentylfuran, Sigma-Aldrich®
2-Pentylfuran
Formula:
C₉H₁₄O Boiling Pt: 64 °C (1013 hPa) Density: 0.838 g/cm³ (19 °C) |
MDL Number:
MFCD00036497 CAS Number: 3777-69-3 EINECS: 223-234-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...