Baicalein, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
M-Clarity™ quality level = MQ100
465119-100MG
465119-500MG
SIAL465119-100MGEA
49.2
EUR
SIAL465119-100MG
SIAL465119-500MG
Baicalein, Sigma-Aldrich®
Baicalein
Formula:
C₁₅H₁₀O₅ MW: 270.24 g/mol |
MDL Number:
MFCD00017459 CAS Number: 491-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...