3,4-Dihydroxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol
M-Clarity™ quality level = MQ200
123447-100G
123447-25G
123447-5G
SIAL123447-100GEA
855
EUR
SIAL123447-100G
SIAL123447-25G
SIAL123447-5G
3,4-Dihydroxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®
3,4-Dihydroxy-3-cyclobutene-1,2-dione
Formula:
C₄H₂O₄ MW: 114.06 g/mol Melting Pt: 293 Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00001334 CAS Number: 2892-51-5 EINECS: 220-761-4 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...