3-Phenylpropionyl chloride
Supplier: Thermo Scientific
SB01152.1
SB01152.10
SB01152.50
MAYBSB01152.1EA
0
EUR
MAYBSB01152.1
MAYBSB01152.10
MAYBSB01152.50
3-Phenylpropionyl chloride
3-Phenylpropionyl chloride
Formula:
C₉H₉ClO Boiling Pt: 232 °C (1013 hPa) Melting Pt: –2 °C Density: 0.997 g/cm³ (20 °C) |
MDL Number:
MFCD00000748 CAS Number: 645-45-4 EINECS: 211-443-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...