4,4,5,5,5-Pentafluoropentan-1-ol

Supplier: Thermo Scientific

Synonyms: 4,4,5,5,5-Pentafluoropentanol

SB01470.1 SB01470.10
MAYBSB01470.1EA 0 EUR
MAYBSB01470.1 MAYBSB01470.10
4,4,5,5,5-Pentafluoropentan-1-ol
4,4,5,5,5-Pentafluoropentan-1-ol
Formula: C₅H₇F₅O
MDL Number: MFCD00153224
CAS Number: 148043-73-6
EINECS: 421-360-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR