4,5-Dicyanoimidazole 99%
Supplier: Thermo Fisher Scientific
Danger
368530050 368530250
ACRO368530050EA 39.7 EUR
ACRO368530050 ACRO368530250
4,5-Dicyanoimidazole 99%
4,5-Dicyanoimidazole
Formula: C₅H₂N₄
Melting Pt: 174…177 °C
Density: 1,458 g/cm³ (20 °C)
MDL Number: MFCD00005194
CAS Number: 1122-28-7
EINECS: 214-344-6
Order Now
Specification Test Results
Appearance White to off-white to light beige Powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
HPLC ≥98.5 %