Genistein, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
M-Clarity™ quality level = MQ200
G6776-10MG
G6776-5MG
SIALG6776-10MGEA
130
EUR
SIALG6776-10MG
SIALG6776-5MG
Genistein, Sigma-Aldrich®
Genistein
Formula:
C₁₅H₁₀O₅ Melting Pt: 297…298 °C |
MDL Number:
MFCD00016952 CAS Number: 446-72-0 EINECS: 207-174-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...