4-Ethylbenzenesulphonyl chloride

Supplier: Thermo Scientific

Synonyms: 4-Ethylbenzene-1-sulphonyl chloride

BTB10241DA BTB10241.10 BTB10241.50
MAYBBTB10241DAEA 0 EUR
MAYBBTB10241DA MAYBBTB10241.10 MAYBBTB10241.50
4-Ethylbenzenesulphonyl chloride
4-Ethylbenzenesulphonyl chloride
Formula: C₈H₉ClO₂S
Boiling Pt: 143 °C (16 mmHg)
Melting Pt: 12 °C
Density: 1.268 g/cm³ (20 °C)
MDL Number: MFCD00041480
CAS Number: 16712-69-9
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR