4-Methylbenzamidine hydrochloride

Supplier: Thermo Scientific

Synonyms: 4-Methylphenylamidine hydrochloride, 4-Methylbenzene-1-carboximidamide hydrochloride, 4-Methylbenzimidamide hydrochloride

SPB07337.1 SPB07337.10
MAYBSPB07337.1EA 0 EUR
MAYBSPB07337.1 MAYBSPB07337.10
4-Methylbenzamidine hydrochloride
4-Methylbenzamidine hydrochloride
Formula: C₈H₁₁ClN₂
MW: 170.64 g/mol
MDL Number: MFCD00040183
CAS Number: 6326-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR