4-Phenoxybenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L17664.03
L17664.06
L17664.03EA
78.4
EUR
L17664.03
L17664.06
4-Phenoxybenzenesulphonyl chloride ≥97%
4-Phenoxybenzenesulphonyl chloride
Purity: 97%
CAS No.: 1623-92-3
Molecular Formula: C12H9ClO3S
Formula Weight: 268.72
Melting Point: 42-45°
Boiling Point: 180°/5mm
UN No.: 3260
MDL No.: MFCD00625748
Formula:
C₁₂H₉ClO₃S Boiling Pt: 180 °C (5 mmHg) Melting Pt: 42…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00625748 CAS Number: 1623-92-3 UN: 3260 ADR: 8,II |