4-Propylbenzenesulphonyl chloride
Supplier: Thermo Scientific
Synonyms:
4-Propylbenzene-1-sulphonyl chloride
BTB10242.1
BTB10242.10
BTB10242.50
MAYBBTB10242.1EA
0
EUR
MAYBBTB10242.1
MAYBBTB10242.10
MAYBBTB10242.50
4-Propylbenzenesulphonyl chloride
4-Propylbenzenesulphonyl chloride
Formula:
C₉H₁₁ClO₂S Boiling Pt: 113…113.5 °C (0.2 mmHg) Melting Pt: –4 °C |
MDL Number:
MFCD00041508 CAS Number: 63014-04-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...