2-(2-Butoxyethoxy)ethyl acetate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Diethyleneglycolmonobutyletheracetate, Diethylene glycol monobutyl ether acetate
A16065.AK
A16065.AU
A16065.AKEA
27.4
EUR
A16065.AK
A16065.AU
2-(2-Butoxyethoxy)ethyl acetate ≥98%
2-(2-Butoxyethoxy)ethyl acetate
Purity: 98%
CAS No.: 124-17-4
Molecular Formula: CH3CO2CH2CH2OCH2CH2O(CH2)3CH3
Formula Weight: 204.27
Flash Point: 115°(239°F)
Physical Form: Liquid
Density: 0.977
Refractive Index: 1.4260
Melting Point: -32°
Boiling Point: 237-241°
Beilstein Reference: 1771533
MDL No.: MFCD00009458
Formula:
C₁₀H₂₀O₄ Boiling Pt: 237…241 °C Melting Pt: –32 °C Density: 0.977 Flash Pt: 115 °C (239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009458 CAS Number: 124-17-4 EINECS: 204-685-9 |