Tributylvinyltin, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Tri-n-butylvinyltin, Tributylvinylstannane
M-Clarity™ quality level = MQ100
271438-1G
271438-25G
271438-5G
SIAL271438-1GEA
64.7
EUR
SIAL271438-1G
SIAL271438-25G
SIAL271438-5G
Tributylvinyltin, Sigma-Aldrich®
Tributylvinyltin
Formula:
C₁₄H₃₀Sn Boiling Pt: 73…74 °C (0.1 mmHg) Density: 1.085 g/cm³ (25 °C) |
MDL Number:
MFCD00009421 CAS Number: 7486-35-3 EINECS: 231-291-4 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...