Hydroxylammonium chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride
M-Clarity™ quality level = MQ200
431362-250G 431362-50G
SIAL431362-250GEA 328 EUR
SIAL431362-250G SIAL431362-50G
Hydroxylammonium chloride, Sigma-Aldrich®
Hydroxylammonium chloride
Formula: NH₂OH·HCl
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853