Lanthanum(III) chloride hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Lanthanum trichloride hydrate
M-Clarity™ quality level = MQ100
211605-100G 211605-500G
SIAL211605-100GEA 109 EUR
SIAL211605-100G SIAL211605-500G
Lanthanum(III) chloride hydrate, Sigma-Aldrich®
Lanthanum (III) chloride hydrate
Formula: Cl₃La·H₂O
MW: 263.28
Boiling Pt: 1000
Melting Pt: 96
Storage Temperature: Ambient
MDL Number: MFCD00149753
CAS Number: 20211-76-1
EINECS: 233-237-5