Butyl methacrylate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methacrylic acid butyl ester
M-Clarity™ quality level = MQ200
235865-100ML 235865-1L 235865-18L
SIAL235865-100MLEA 58.5 EUR
SIAL235865-100ML SIAL235865-1L SIAL235865-18L
Butyl methacrylate, Sigma-Aldrich®
Butyl methacrylate
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 163
Melting Pt: –75 °C
Density: 0.8951 g/cm³ (20 °C)
MDL Number: MFCD00009444
CAS Number: 97-88-1
EINECS: 202-615-1
UN: 2227
ADR: 3 III