Sodium ethoxide, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Sodium ethylate, Sodium ethanolate

M-Clarity™ quality level = MQ200

156248-100G 156248-500G 156248-5G
SIAL156248-100GEA 61.5 EUR
SIAL156248-100G SIAL156248-500G SIAL156248-5G
Sodium ethoxide, Sigma-Aldrich®
Sodium ethoxide
Formula: C₂H₅NaO
Melting Pt: 260 °C
Density: 0.868 g/cm³ (20 °C)
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR