Sodium triacetoxyborohydride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

316393-100G 316393-1KG 316393-25G
SIAL316393-100GEA 136 EUR
SIAL316393-100G SIAL316393-1KG SIAL316393-25G
Sodium triacetoxyborohydride, Sigma-Aldrich®
Sodium triacetoxyborohydride
Formula: C₆H₁₀BNaO₆
Melting Pt: 114…118 °C
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 3396
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR