Acrylamide : Bisacrylamide (19:1)
Supplier: Ambion
Danger
9022 9024
AMBI9022EA 0 EUR
AMBI9022 AMBI9024
Acrylamide : Bisacrylamide (19:1)
Acrylamide-Bis (32:1)
Storage Temperature: Refrigerator
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1,III