2-Pentadecanone ≥97%
Supplier: Thermo Fisher Scientific
L09347.14
L09347.22
L09347.14EA
50
EUR
L09347.14
L09347.22
2-Pentadecanone ≥97%
2-Pentadecanone
Purity: 97%
CAS No.: 2345-28-0
Molecular Formula: C15H30O
Formula Weight: 226.40
Flash Point: 144°(291°F)
Melting Point: 37-41°
Boiling Point: 293-294°
Beilstein Reference: 1764872
MDL No.: MFCD00053712
Formula:
C₁₅H₃₀O Boiling Pt: 293…294 °C Melting Pt: 37…41 °C Density: 0.82 Flash Pt: 144 °C (291 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053712 CAS Number: 2345-28-0 EINECS: 219-064-8 |