Bottles, wide neck, square with thread
Supplier: MAUSER
3005028 3005620 3006593 3010631 3011615 3021112 3040025 3005015 3005025 3005633 3006603 3010644 3011630 3021125 3040012 3041024 81600076 81600074 81600077 81600154
215-2830EA 247 EUR
215-2830 215-2831 215-2833 215-2835 215-2837 215-2839 215-2842 215-2828 215-2829 215-2832 215-2834 215-2836 215-2838 215-2840 215-2841 215-2843 215-3292 215-3291 215-3293 215-3294
Bottles, wide neck, square with thread
Bottles Plastic Bottles
HDPE, translucent or HDPE, white.
  • Good chemical resistance
  • Resistant to temperatures from −40 to +110 °C
  • Tamper-evident closures must be ordered separately (see below for available options)
Ideal for collecting, storing, and shipping liquids, pastes, and granulates. Square shape saves space, recessed labelling space. Wide neck makes filling and cleaning easy.