Watch glasses
Supplier: Brand
150010 150015 150020 150030 150040 150045 150050 150060 150065 150070 150080
BRND150010EA 27 EUR
BRND150010 BRND150015 BRND150020 BRND150030 BRND150040 BRND150045 BRND150050 BRND150060 BRND150065 BRND150070 BRND150080
Watch glasses
Watch Glasses
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.