Hydroxyacetone 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
L15008.22
L15008.36
L15008.22EA
31.2
EUR
L15008.22
L15008.36
Hydroxyacetone 95%
Hydroxyacetone
Purity: 95%
CAS No.: 116-09-6
Molecular Formula: C3H6O2
Formula Weight: 74.08
Storage Temperature: 2°C to 8°C
Flash Point: 56°(132°F)
Refractive Index: 1.4250
Odour: Characteristic
pH: 3.4
Melting Point: -17°
Boiling Point: 145-146°
Beilstein Reference: 605368
Merck Reference: 14,65
UN No.: 1224
MDL No.: MFCD00004669
Formula:
C₃H₆O₂ Boiling Pt: 145…146 °C Melting Pt: –17 °C Density: 1.080 Flash Pt: 56 °C (132 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004669 CAS Number: 116-09-6 EINECS: 204-124-8 UN: 1224 ADR: 3,III Merck Index: 14,65 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...