Acetylacetone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: ACAC, 2,4-Pentanedione
10916-185KG 10916-1KG 10916-25KG
SIAL10916-185KGEA 3890 EUR
SIAL10916-185KG SIAL10916-1KG SIAL10916-25KG
Acetylacetone, Sigma-Aldrich®
Acetylacetone
Packaging: produced by Wacker Chemie AG
Formula: CH₃COCH₂COCH₃
MW: 100.12 g/mol
Boiling Pt: 140.4
Melting Pt: –23 °C
Density: 0.96945 g/cm³ (20 °C)
Flash Pt: 30
Storage Temperature: Ambient
MDL Number: MFCD00008787
CAS Number: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3 III
Merck Index: 13,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c85a64bf9c7997e9ab03e421a93e7c71e57b8a3&vid=134ed3029693deef3759692be256c57b3a58b087&prid=web&clid=SAPDAM