Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Warning
Synonyms: Methylene chloride
Stabilization: 0.1 % Stabilised with ethanol
Filtered through a 0.2 µm filter, packaged under nitrogen.
83623.320EA 154 EUR
83623.320 83623.290
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.3300 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (GC) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 10 ppm
Water Max. 0.05 %
Absorbance (240 nm) Max. 0.3
Absorbance (245 nm) Max. 0.1
Absorbance (260 nm) Max. 0.01
Transmittance (240 nm) Min. 50 %
Transmittance (245 nm) Min. 80 %
Transmittance (260 nm) Min. 98 %