3-Chloro-1-propanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Chloropropan-1-ol, 1-Chloro-3-hydroxypropane
M-Clarity™ quality level = MQ200
C46403-25G C46403-100G C46403-500G
SIALC46403-25GEA 47.9 EUR
SIALC46403-25G SIALC46403-100G SIALC46403-500G
3-Chloro-1-propanol, Sigma-Aldrich®
3-Chloro-1-propanol
Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 161
Melting Pt: –20 °C
Density: 1.1309 g/cm³ (20 °C)
Flash Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00002943
CAS Number: 627-30-5
EINECS: 210-992-9
UN: 2849
ADR: 6.1 III