Trifluoroacetic acid
Supplier: Thermo Fisher Scientific
Danger
28903 28904 28901 28902
PIER28903EA 249 EUR
PIER28903 PIER28904 PIER28901 PIER28902
Trifluoroacetic acid
Trifluoroacetic acid
Formula: C₂HF₃O₂
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM