1-Iodododecane 98% stabilised

Supplier: Thermo Fisher Scientific
Warning


Stabilization: Stabilised with silver

250090250 250091000 250095000
ACRO250090250EA 44.3 EUR
ACRO250090250 ACRO250091000 ACRO250095000
1-Iodododecane 98% stabilised
1-Iodododecane
Formula: C₁₂H₂₅I
Boiling Pt: 159…160 °C (15 mmHg)
Melting Pt: –3 °C
Density: 1.2 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001088
CAS Number: 4292-19-7
EINECS: 224-293-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4825 to 1.4865 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Copper

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR