1-Iodopropane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Propyl iodide
M-Clarity™ quality level = MQ100
171883-100G 171883-500G 171883-5G
SIAL171883-100GEA 57.6 EUR
SIAL171883-100G SIAL171883-500G SIAL171883-5G
1-Iodopropane, Sigma-Aldrich®
1-Iodopropane
Formula: C₃H₇I
MW: 169.99 g/mol
Boiling Pt: 103
Melting Pt: –98 °C
Density: 1.741 g/cm³ (20 °C)
Flash Pt: 44
Storage Temperature: Ambient
MDL Number: MFCD00001095
CAS Number: 107-08-4
EINECS: 203-460-2
UN: 2392
ADR: 3 III
Merck Index: 12,08047