Triethylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (Diethylamino)ethane, TEA
90337-100ML 90337-25ML 90337-5ML
SIAL90337-100MLEA 808 EUR
SIAL90337-100ML SIAL90337-25ML SIAL90337-5ML
Triethylamine, Sigma-Aldrich®
Triethylamine
Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM