Tripropylamine 98%
Supplier: Thermo Fisher Scientific
Danger
162690010 162690025
ACRO162690010EA 99.3 EUR
ACRO162690010 ACRO162690025
Tripropylamine 98%
Tripropylamine
Formula: C₉H₂₁N
MW: 143.27 g/mol
Boiling Pt: 156 °C (760 mmHg)
Melting Pt: –93 °C
Density: 0.76 g/cm³
Flash Pt: 32.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00009363
CAS Number: 102-69-2
EINECS: 203-047-7
UN: 2260
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4155 to 1.4175 (20°C, 589 nm)
Water ≤0.2 %
GC ≤0.2 % dipropylamine