Ethyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
B25180.06 B25180.14
B25180.06EA 42.6 EUR
B25180.06 B25180.14
Ethyl isocyanate ≥98%
Ethyl isocyanate
Formula: C₃H₅NO
Boiling Pt: 60 °C (1013 hPa)
Melting Pt: < –50 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 109-90-0
EINECS: 203-717-9
UN: 2481
ADR: 3,I