Formaldehyde diethyl acetal, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethylal, Diethoxymethane
47675-100ML-F 47675-500ML-F
SIAL47675-100ML-FEA 69.4 EUR
SIAL47675-100ML-F SIAL47675-500ML-F
Formaldehyde diethyl acetal, Sigma-Aldrich®
Formaldehyde diethyl acetal
Formula: C₅H₁₂O₂
Boiling Pt: 88 °C (1013 hPa)
Melting Pt: –66 °C
Density: 0.8319 g/cm³ (20 °C)
Flash Pt: –6 °C
Storage Temperature: Ambient
MDL Number: MFCD00009246
CAS Number: 462-95-3
EINECS: 207-330-6
UN: 2373
ADR: 3,II